Proteintech
Manufacturer Code:160641AP150UL
Catalog # 50-173-0687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PMPCB Fusion Protein Ag8937 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta MPP MPP11 MPPB MPPP52 P 52 PMPCB; Beta-MPP; mitochondrial processing peptidase beta subunit; Mitochondrial-processing peptidase subunit beta; P-52; peptidase (mitochondrial processing) beta
Gene Aliases: 3110004O18Rik; Beta-MPP; MPP11; MPPB; MPPP52; P-52; PMPCB
UniProt ID: (Human) O75439, (Rat) Q03346, (Mouse) Q9CXT8
Entrez Gene ID: (Human) 9512, (Rat) 64198, (Mouse) 73078
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.