Novus Biologicals
Manufacturer Code:NBP155475
Catalog # NBP155475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PMM1/Phosphomannomutase 1. The peptide sequence was selected from the N terminal of PMM1/Phosphomannomutase 1. Peptide sequence MAVTAQAARRKERVLCLFDVDGTLTPARQKIDPEVAAFLQKLRSRVQIGV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain glucose-1,6-bisphosphatase; brain glucose-16-bisphosphatase EC 5.4.2.8 phosphomannomutase 1 PMM 1 PMMH22 PMMH-22 Sec53; Phosphomannomutase 1; PMM 1; PMMH-22
Gene Aliases: PMM1; PMMH22; Sec53
UniProt ID: (Human) Q92871
Entrez Gene ID: (Human) 5372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.