Invitrogen
Manufacturer Code:PA556547
Catalog # PIPA556547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PMM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain glucose-1,6-bisphosphatase; Phosphomannomutase 1; PMM 1; PMMH-22; Secp53 homolog
Gene Aliases: C77612; PMM1; PMMH22; Sec53
UniProt ID: (Human) Q92871, (Mouse) O35621
Entrez Gene ID: (Human) 5372, (Mouse) 29858, (Rat) 300089
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.