Invitrogen
Manufacturer Code:PA5112656
Catalog # PIPA5112656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosphomannomutase 1 protein (2-262AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain glucose-1,6-bisphosphatase; brain glucose-16-bisphosphatase C77612 Phosphomannomutase 1 PMM 1 PMM1 PMMH22 PMMH-22 Sec53 Secp53 homolog; Phosphomannomutase 1; PMM 1; PMMH-22; Secp53 homolog
Gene Aliases: C77612; PMM1; PMMH22; Sec53
UniProt ID: (Human) Q92871, (Mouse) O35621
Entrez Gene ID: (Human) 5372, (Mouse) 29858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.