Novus Biologicals
Manufacturer Code:NB11082398
Catalog # NB11082398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from Nter domain of mouse PML protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: MYLPP8675 promyelocytic leukemia Promyelocytic leukemia protein RING finger protein 71 RNF71probable transcription factor PML TRIM19promyelocytic leukemia inducer of tripartite motif protein TRIM19 Tripartite motif-containing protein 19; PML/RARA fusion; probable transcription factor PML; Promyelocytic leukemia protein; promyelocytic leukemia, inducer of; Protein PML; RING finger protein 71; tripartite motif protein TRIM19; Tripartite motif-containing protein 19
Gene Aliases: MYL; PML; PP8675; RNF71; TRIM19
UniProt ID: (Human) Q9BPW2
Entrez Gene ID: (Human) 5371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.