Invitrogen
Manufacturer Code:PA579835
Catalog # PIPA579835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human PML Protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PML/RARA fusion; probable transcription factor PML; promyelocytic leukemia; Promyelocytic leukemia protein; promyelocytic leukemia, inducer of; Protein PML; Protein PML Promyelocytic leukemia protein RING finger protein 71 Tripartite motif-containing protein 19 PML MYL PP8675 RNF71 TRIM19; RING finger protein 71; tripartite motif protein TRIM19; Tripartite motif-containing protein 19
Gene Aliases: 1200009E24Rik; AI661194; MYL; PML; PP8675; RGD1562602; RNF71; TRIM19
UniProt ID: (Human) Q9BPW2, (Mouse) Q60953
Entrez Gene ID: (Human) 5371, (Mouse) 18854, (Rat) 315713
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.