Invitrogen
Manufacturer Code:PA529614
Catalog # PIPA529614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 233 of Human PML |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MYL PP8675 RNF71 TRIM19 RING finger protein 71 probable transcription factor PML promyelocytic leukemia protein promyelocytic leukemia inducer of protein PML tripartite motif protein TRIM19 tripartite motif-containing protein 19; PML/RARA fusion; probable transcription factor PML; Promyelocytic leukemia protein; promyelocytic leukemia, inducer of; Protein PML; RING finger protein 71; tripartite motif protein TRIM19; Tripartite motif-containing protein 19
Gene Aliases: MYL; PML; PP8675; RNF71; TRIM19
UniProt ID: (Human) Q9BPW2
Entrez Gene ID: (Human) 5371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.