Abnova
Manufacturer Code:PAB24780
Catalog # 89-126-448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to PML. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PML.
For Research Use Only
Protein Aliases: MYL; PML/RARA fusion; PP8675; probable transcription factor PML; Promyelocytic leukemia protein; promyelocytic leukemia, inducer of; Protein PML; RING finger protein 71; RNF71; TRIM19; tripartite motif protein TRIM19; Tripartite motif-containing protein 19
Gene Aliases: MYL; PML; PP8675; RNF71; TRIM19
UniProt ID: (Human) Q9BPW2
Entrez Gene ID: (Human) 5371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.