Novus Biologicals
Manufacturer Code:NBP169571
Catalog # NBP169571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SILV(silver homolog (mouse)) The peptide sequence was selected from the N terminal of human PMEL (NP_008859). Peptide sequence HFLRNQPLTFALQLHDPSGYLAEADLSYTWDFGDSSGTLISRALVVTHTY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 95 kDa melanocyte-specific secreted glycoprotein; D12S53EP1 gp100 ME20 ME20-M melanocyte protein mel 17 Melanocyte protein Pmel 17 Melanocytes lineage-specific antigen GP100 Melanoma-associated ME20 antigen melanosomal matrix protein17 PMEL17P100 premelanosome proteinME20M SI SIL silver (mouse homolog) like silver homolog (mouse) Silver locus protein homolog silver mouse homolog of SILVPmel17; M-alpha; M-beta; ME20-M; ME20-S; ME20M; melanocyte protein mel 17; Melanocyte protein PMEL; Melanocyte protein Pmel 17; Melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; P26; Premelanosome protein; Secreted melanoma-associated ME20 antigen; Silver locus protein homolog; silver, mouse, homolog of
Gene Aliases: D12S53E; gp100; ME20; ME20-M; ME20M; P1; P100; PMEL; PMEL17; SI; SIL; SILV
UniProt ID: (Human) P40967
Entrez Gene ID: (Human) 6490
Molecular Function: cell adhesion molecule membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.