Invitrogen
Manufacturer Code:PA5111763
Catalog # PIPA5111763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 601-617 of rat PMCA4 (2nd intracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP2B2 Atp2b4 BOS_15586 Matrix-remodeling-associated protein 1 MXRA1 Plasma membrane calcium ATPase isoform 4 Plasma membrane calcium pump isoform 4 Plasma membrane calcium-transporting ATPase 4 PMCA4 PMCA4b PMCA4x; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 4; PMCA4
Gene Aliases: Atp2b4; PMCA4
UniProt ID: (Rat) Q64542
Entrez Gene ID: (Rat) 29600
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.