Novus Biologicals
Manufacturer Code:NBP197754
Catalog # NBP197754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KQDGAIENRNKAKAQD corresponding to internal region according to NP_001673.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase Ca++ transporting plasma membrane 1 EC 3.6.3 EC 3.6.3.8 plasma membrane calcium ATPase Plasma membrane calcium ATPase isoform 1 plasma membrane calcium pump Plasma membrane calcium pump isoform 1 plasma membrane calcium-ATPase PMCA1kb PMCA1plasma membrane calcium-transporting ATPase 1; ATPase, Ca++ transporting, plasma membrane 1; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium-transporting ATPase 1; PMCA1
Gene Aliases: ATP2B1; PMCA1; PMCA1kb
UniProt ID: (Human) P20020
Entrez Gene ID: (Human) 490
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.