Invitrogen
Manufacturer Code:PA5114932
Catalog # PIPA5114932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Calcium Pump PMCA4 ATPase corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810442I22Rik 6430519O13Rik ATP2B ATP2B1 Atp2b2 ATP2B3 ATP2B4 ATPase isoform 2 Na+K+ transporting beta polypeptide 2 ATPase isoform 2 Na+K+ transporting beta polypeptide 2 ATPase plasma membrane Ca2+ transporting 1 ATPase plasma membrane Ca2+ transporting 2 ATPase plasma membrane Ca2+ transporting 3 ATPase plasma membrane Ca2+ transporting 4 ATPase Ca++ transporting plasma membrane 1 ATPase Ca++ transporting plasma membrane 2 ATPase Ca++ transporting plasma membrane 3 ATPase Ca++ transporting plasma membrane 4 Ca2+/Mg2+ ATPase CFAP39 cilia and flagella associated protein 39 CLA2 D6Abb2e deaf waddler dfw E130111D10Rik Gena300 jog Matrix-remodeling-associated protein 1 MXRA1 OPCA plasma membrane Ca(2+)-ATPase plasma membrane Ca++-ATPase plasma membrane Ca2+ pump 2 plasma membrane Ca2+-ATPases 4 plasma membrane calcium ATPase Plasma membrane calcium ATPase isoform 1 Plasma membrane calcium ATPase isoform 2 Plasma membrane calcium ATPase isoform 3 Plasma membrane calcium ATPase isoform 4 plasma membrane calcium pump Plasma membrane calcium pump isoform 1 Plasma membrane calcium pump isoform 2 Plasma membrane calcium pump isoform 3 Plasma membrane calcium pump isoform 4 Plasma membrane calcium-transporting ATPase 1 plasma membrane calcium-transporting ATPase 2 Plasma membrane calcium-transporting ATPase 3 plasma membrane calcium-transporting ATPase 4 PMCA PMCA1 Pmca1a Pmca1b Pmca1c PMCA1kb PMCA2 PMCA2a PMCA2i Pmca3 PMCA3a Pmca4 PMCA4b PMCA4x sarcolemmal calcium pump SCAX1 Tmy wms wri wriggle; ATPase isoform 2 Na+K+ transporting beta polypeptide 2; ATPase isoform 2, Na+K+ transporting, beta polypeptide 2; ATPase, Ca++ transporting, plasma membrane 1; ATPase, Ca++ transporting, plasma membrane 2; ATPase, Ca++ transporting, plasma membrane 3; ATPase, Ca++ transporting, plasma membrane 4; cilia and flagella associated protein 39; deaf waddler; Matrix-remodeling-associated protein 1; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 1; Plasma membrane calcium-transporting ATPase 2; Plasma membrane calcium-transporting ATPase 3; Plasma membrane calcium-transporting ATPase 4; PMCA1; PMCA2; PMCA3; PMCA4; sarcolemmal calcium pump; wriggle
Gene Aliases: 2810442I22Rik; 6430519O13Rik; ATP2B1; ATP2B2; ATP2B3; ATP2B4; CFAP39; CLA2; D6Abb2e; dfw; E130111D10Rik; Gena300; jog; MXRA1; OPCA; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb; PMCA2; PMCA2a; PMCA2i; PMCA3; PMCA3a; PMCA4; PMCA4b; PMCA4x; SCAX1; Tmy; wms; wri
UniProt ID: (Human) P20020, (Human) Q01814, (Human) Q16720, (Human) P23634, (Mouse) G5E829, (Rat) P11505, (Rat) P11506, (Mouse) Q9R0K7, (Rat) Q64568, (Rat) Q64542, (Mouse) Q6Q477
Entrez Gene ID: (Human) 490, (Human) 491, (Human) 492, (Human) 493, (Mouse) 67972, (Rat) 29598, (Rat) 24215, (Mouse) 11941, (Rat) 29599, (Mouse) 320707, (Rat) 29600, (Mouse) 381290
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.