Invitrogen
Manufacturer Code:PA5113788
Catalog # PIPA5113788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human ATP2B3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810442I22Rik 6430519O13Rik ATP2B Atp2b1 Atp2b2 ATP2B3 Atp2b4 ATPase isoform 2 Na+K+ transporting beta polypeptide 2 ATPase isoform 2 Na+K+ transporting beta polypeptide 2 ATPase plasma membrane Ca2+ transporting 1 ATPase plasma membrane Ca2+ transporting 2 ATPase plasma membrane Ca2+ transporting 3 ATPase plasma membrane Ca2+ transporting 4 ATPase Ca++ transporting plasma membrane 1 ATPase Ca++ transporting plasma membrane 2 ATPase Ca++ transporting plasma membrane 3 ATPase Ca++ transporting plasma membrane 4 Ca2+/Mg2+ ATPase CFAP39 cilia and flagella associated protein 39 CLA2 D6Abb2e deaf waddler dfw E130111D10Rik Gena300 jog Matrix-remodeling-associated protein 1 MXRA1 OPCA plasma membrane Ca(2+)-ATPase plasma membrane Ca++-ATPase plasma membrane Ca2+ pump 2 plasma membrane Ca2+-ATPases 4 plasma membrane calcium ATPase Plasma membrane calcium ATPase isoform 1 Plasma membrane calcium ATPase isoform 2 Plasma membrane calcium ATPase isoform 3 Plasma membrane calcium ATPase isoform 4 plasma membrane calcium pump Plasma membrane calcium pump isoform 1 Plasma membrane calcium pump isoform 2 Plasma membrane calcium pump isoform 3 Plasma membrane calcium pump isoform 4 Plasma membrane calcium-transporting ATPase 1 Plasma membrane calcium-transporting ATPase 2 Plasma membrane calcium-transporting ATPase 3 Plasma membrane calcium-transporting ATPase 4 PMCA PMCA1 Pmca1a Pmca1b Pmca1c PMCA1kb Pmca2 PMCA2a PMCA2i Pmca3 PMCA3a PMCA4 PMCA4b PMCA4x sarcolemmal calcium pump SCAX1 Tmy wms wri wriggle; ATPase, Ca++ transporting, plasma membrane 1; ATPase, Ca++ transporting, plasma membrane 2; ATPase, Ca++ transporting, plasma membrane 3; ATPase, Ca++ transporting, plasma membrane 4; cilia and flagella associated protein 39; Matrix-remodeling-associated protein 1; plasma membrane Ca(2+)-ATPase; plasma membrane Ca2+ pump 2; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 1; Plasma membrane calcium-transporting ATPase 2; Plasma membrane calcium-transporting ATPase 3; Plasma membrane calcium-transporting ATPase 4; PMCA1; PMCA2; PMCA3; PMCA4; sarcolemmal calcium pump
Gene Aliases: ATP2B1; ATP2B2; ATP2B3; ATP2B4; CFAP39; CLA2; MXRA1; OPCA; PMCA1; PMCA1kb; PMCA2; PMCA2a; PMCA2i; PMCA3; PMCA3a; PMCA4; PMCA4b; PMCA4x; SCAX1
UniProt ID: (Human) P20020, (Human) Q01814, (Human) Q16720, (Human) P23634
Entrez Gene ID: (Human) 490, (Human) 491, (Human) 492, (Human) 493
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.