Invitrogen
Manufacturer Code:PA5107073
Catalog # PIPA5107073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: FAGEKFFDID SGRKAPLHSP PSQHYTIVFN TFVLMQLFNE INSRKIHGEK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase, Ca++ transporting, plasma membrane 4; Matrix-remodeling-associated protein 1; Plasma membrane calcium ATPase isoform 4; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 4; PMCA4; sarcolemmal calcium pump
Gene Aliases: ATP2B2; ATP2B4; MXRA1; PMCA4; PMCA4b; PMCA4x
UniProt ID: (Human) P23634
Entrez Gene ID: (Human) 493
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.