Invitrogen
Manufacturer Code:PA511229
Catalog # PIPA511229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 452-481 amino acids from the C-terminal region of human PMAT(Slc29a4) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ENT4 PMAT FLJ34923 Solute carrier family 29 (Nucleoside transporters) member 4; Equilibrative nucleoside transporter 4; hENT4; Plasma membrane monoamine transporter; solute carrier family 29 (equilibrative nucleoside transporter), member 4; solute carrier family 29 (nucleoside transporters), member 4; Solute carrier family 29 member 4
Gene Aliases: ENT4; PMAT; PSEC0113; SLC29A4
UniProt ID: (Human) Q7RTT9
Entrez Gene ID: (Human) 222962
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.