R&D Systems
Manufacturer Code:MAB2944SP
Catalog # MAB2944SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6318100 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PLZF Met1-Gln254 Accession # Q05516 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PLZFzinc finger and BTB domain-containing protein 16 ZBTB16 zinc finger and BTB domain containing 16 Zinc finger protein 145 zinc finger protein 145 (Kruppel-like expressed in promyelocytic leukemia) Zinc finger protein PLZF ZNF145 ZNF145Promyelocytic leukemia zinc finger protein; promyelocytic leukaemia zinc finger; Promyelocytic leukemia zinc finger protein; Zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); Zinc finger protein PLZF
Gene Aliases: PLZF; ZBTB16; ZNF145
UniProt ID: (Human) Q05516
Entrez Gene ID: (Human) 7704
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.