Invitrogen
Manufacturer Code:PA548065
Catalog # PIPA548065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit human Plexin B3 His45-Gln1255 (Glu1156Asp) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: PLEXB3 PLEXR PLXN6 PLXNB3 PLEXB3 plexin B3 plexin-B3 PLEXR; plexin 6; Plexin-B3
Gene Aliases: KIAA1206; PLEXB3; PLEXR; PLXN6; PLXNB3
UniProt ID: (Human) Q9ULL4
Entrez Gene ID: (Human) 5365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.