Invitrogen
Manufacturer Code:PA547882
Catalog # PIPA547882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Plexin B3 His25-Gln1235 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: PLEXB3 PLEXR PLXN6 PLXNB3 PLEXB3 plexin B3 plexin-B3 PLEXR; plexin 6; Plexin-6; Plexin-B3; plexin-B3 {ECO:0000303|PubMed:20696765}; plxnb3 {ECO:0000312|Ensembl:ENSRNOP00000053235}; Protein Plxnb3
Gene Aliases: AI451018; Kiaa1206; Plxn6; Plxnb3; RGD1560615
UniProt ID: (Mouse) Q9QY40, (Rat) D3ZLH5
Entrez Gene ID: (Mouse) 140571, (Rat) 363517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.