Invitrogen
Manufacturer Code:PA545692
Catalog # PIPA545692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PLXNB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: plexin 6; plexin B3 FLJ76953 PLEXB3 PLEXR PLXN6; Plexin-B3
Gene Aliases: KIAA1206; PLEXB3; PLEXR; PLXN6; PLXNB3
UniProt ID: (Human) Q9ULL4
Entrez Gene ID: (Human) 5365, (Guinea pig) 100712880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.