Proteintech
Manufacturer Code:237951AP150UL
Catalog # 50-173-4127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLXNB1 Fusion Protein Ag20719 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0407 plexin B1 PLXN5 PLXNB1 Semaphorin receptor SEP SEP; plexin 5; Plexin-B1; Semaphorin receptor SEP
Gene Aliases: 2900002G15Rik; AU042020; KIAA0407; mKIAA0407; PLEXIN-B1; PLXN5; PLXNB1; SEP
UniProt ID: (Human) O43157, (Mouse) Q8CJH3
Entrez Gene ID: (Human) 5364, (Mouse) 235611
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.