Invitrogen
Manufacturer Code:MA514795
Catalog # PIMA514795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | F.566.2 |
Immunogen | GST-fusion protein corresponding to residues from the human sequence of Plexin A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: OTTHUMP00000207245 plexin-A4; Plexin-A4
Gene Aliases: 9330117B14; FAYV2820; KIAA1550; mKIAA1550; PLEXA4; Plxa4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2, (Mouse) Q80UG2
Entrez Gene ID: (Human) 91584, (Mouse) 243743, (Rat) 312213
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
signaling molecule
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.