Invitrogen
Manufacturer Code:MA524326
Catalog # PIMA524326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 707206 |
Immunogen | CHO-derived recombit human Plexin A4 Thr24-Pro1237 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FAYV2820 PLEXA4 PLXNA4A PLXNA4B PRO34003 plexin-A4; Plexin-A4
Gene Aliases: FAYV2820; KIAA1550; PLEXA4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2
Entrez Gene ID: (Human) 91584
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
signaling molecule
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.