Invitrogen
Manufacturer Code:PA5115772
Catalog # PIPA5115772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Plexin A4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9330117B14 FAYV2820 KIAA1550 mKIAA1550 OTTHUMP00000207245 PLEXA4 plexin A4 plexin A4 A plexin-A4 Plxa4 Plxna4 Plxna4a PLXNA4B PRO34003 UNQ2820/PRO34003; Plexin-A4
Gene Aliases: 9330117B14; FAYV2820; KIAA1550; mKIAA1550; PLEXA4; Plxa4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2, (Mouse) Q80UG2
Entrez Gene ID: (Human) 91584, (Mouse) 243743, (Rat) 312213
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
signaling molecule
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.