Abnova
Manufacturer Code:PAB22749
Catalog # 89-124-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PLXNA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PLXNA4.
For Research Use Only
Protein Aliases: DKFZp434G0625; DKFZp566O0546; FAYV2820; FLJ35026; FLJ38287; KIAA1550; PLEXA4; Plexin-A4; PLXNA4A; PLXNA4B; PRO34003
Gene Aliases: FAYV2820; KIAA1550; PLEXA4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2
Entrez Gene ID: (Human) 91584
Molecular Function: kinase protein kinase protein kinase receptor receptor signaling molecule transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.