Invitrogen
Manufacturer Code:MA523949
Catalog # PIMA523949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 251512 |
Immunogen | E. coli-derived recombit human PLUNC Gln20-Val256 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BPI fold containing family A, member 1; BPI fold-containing family A member 1; BPIFA1 LUNX SPLUNC1 bA49G10.5 ligand-binding protein RYA3 LPLUNC3 Lung-specific protein X LUNXNASG Nasopharyngeal carcinoma-related protein Palate lung and nasal epithelium clone protein palate lung and nasal epithelium associated palate lung and nasal epithelium carcinoma associated protein Plunc Secretory protein in upper respiratory tracts SPLUNC1SPURT tracheal epithelium enriched protein Tracheal epithelium-enriched protein Von Ebner protein Hl; ligand-binding protein RYA3; Lung-specific protein X; Nasopharyngeal carcinoma-related protein; Palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; protein Plunc; Secretory protein in upper respiratory tracts; Short PLUNC1; SPLUNC1; tracheal epithelium enriched protein; Tracheal epithelium-enriched protein; Von Ebner protein Hl
Gene Aliases: bA49G10.5; BPIFA1; LUNX; NASG; PLUNC; SPLUNC1; SPURT; UNQ787/PRO1606
UniProt ID: (Human) Q9NP55
Entrez Gene ID: (Human) 51297
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.