Invitrogen
Manufacturer Code:PA599874
Catalog # PIPA599874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PLUNC corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA49G10.5 BPI fold containing family A member 1 BPI fold containing family A member 1 BPI fold-containing family A member 1 Bpifa1 ES cell-related protein ligand-binding protein RYA3 LPLUNC3 lung-specific protein X LUNX LUNXNASG NASG Nasopharyngeal carcinoma-related protein palate lung and nasal epithelium clone protein palate lung and nasal epithelium associated palate lung and nasal epithelium associated palate lung and nasal epithelium carcinoma associated protein palate lung and nasal epithelium clone palate lung and nasal epithelium expressed transcript Plunc protein Plunc secretory protein in upper respiratory tracts short palate Short PLUNC1 SPLUNC1 SPLUNC1SPURT SPURT tracheal epithelium enriched protein tracheal epithelium-enriched protein UNQ787/PRO1606 Von Ebner protein Hl; BPI fold containing family A, member 1; BPI fold-containing family A member 1; ES cell-related protein; ligand-binding protein RYA3; Lung-specific protein X; Nasopharyngeal carcinoma-related protein; Palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; palate, lung, and nasal epithelium carcinoma associated protein; palate, lung, and nasal epithelium expressed transcript; protein Plunc; Secretory protein in upper respiratory tracts; Short PLUNC1; SPLUNC1; tracheal epithelium enriched protein; Tracheal epithelium-enriched protein; Von Ebner protein Hl
Gene Aliases: bA49G10.5; BPIFA1; LUNX; NASG; PLUNC; SPLUNC1; SPURT; UNQ787/PRO1606
UniProt ID: (Human) Q9NP55, (Rat) Q8K4I4
Entrez Gene ID: (Human) 51297, (Rat) 246238
Molecular Function: antibacterial response protein defense/immunity protein transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.