Novus Biologicals
Manufacturer Code:NBP226120
Catalog # NBP226120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-NEVLRGLDITLVHD corresponding to C-Terminus according to NP_057667.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bA49G10.5 ligand-binding protein RYA3 LPLUNC3 Lung-specific protein X LUNXNASG Nasopharyngeal carcinoma-related protein Palate lung and nasal epithelium clone protein palate lung and nasal epithelium associated palate lung and nasal epithelium carcinoma associated protein Plunc Secretory protein in upper respiratory tracts SPLUNC1SPURT tracheal epithelium enriched protein Tracheal epithelium-enriched protein Von Ebner protein Hl; BPI fold containing family A, member 1; BPI fold-containing family A member 1; ligand-binding protein RYA3; Lung-specific protein X; Nasopharyngeal carcinoma-related protein; Palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; protein Plunc; Secretory protein in upper respiratory tracts; Short PLUNC1; SPLUNC1; tracheal epithelium enriched protein; Tracheal epithelium-enriched protein; Von Ebner protein Hl
Gene Aliases: bA49G10.5; BPIFA1; LUNX; NASG; PLUNC; SPLUNC1; SPURT; UNQ787/PRO1606
UniProt ID: (Human) Q9NP55
Entrez Gene ID: (Human) 51297
Molecular Function: antibacterial response protein defense/immunity protein transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.