R&D Systems
Manufacturer Code:BAF1897
Catalog # BAF1897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PLUNC Gln20-Val256 Accession # Q9NP55 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bA49G10.5 BPIFA1 ligand-binding protein RYA3 LPLUNC3 Lung-specific protein X LUNX LUNXNASG Nasopharyngeal carcinoma-related protein Palate lung and nasal epithelium clone protein palate lung and nasal epithelium associated palate lung and nasal epithelium carcinoma associated protein Plunc Secretory protein in upper respiratory tracts SPLUNC1 SPLUNC1SPURT tracheal epithelium enriched protein Tracheal epithelium-enriched protein Von Ebner protein Hl; BPI fold containing family A, member 1; BPI fold-containing family A member 1; ligand-binding protein RYA3; Lung-specific protein X; Nasopharyngeal carcinoma-related protein; Palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; protein Plunc; Secretory protein in upper respiratory tracts; Short PLUNC1; SPLUNC1; tracheal epithelium enriched protein; Tracheal epithelium-enriched protein; Von Ebner protein Hl
Gene Aliases: bA49G10.5; BPIFA1; LUNX; NASG; PLUNC; SPLUNC1; SPURT; UNQ787/PRO1606
UniProt ID: (Human) Q9NP55
Entrez Gene ID: (Human) 51297
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.