Invitrogen
Manufacturer Code:MA528630
Catalog # PIMA528630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13A6 &lsqbTEC-23] |
Immunogen | Pooled lipid rafts isolated from RBL-2H3 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ca(2+)-dependent phospholipid scramblase 1; Mg(2+)-dependent nuclease; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1
Gene Aliases: Plscr1
UniProt ID: (Rat) P58195
Entrez Gene ID: (Rat) 117540
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.