Proteintech
Manufacturer Code:115821AP150UL
Catalog # 50-172-7285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLSCR1 Fusion Protein Ag2172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; Mg(2+)-dependent nuclease; MmTRA1b; MMTRA1B phospholipid scramblase 1 PL scramblase 1 PLSCR1; NOR1; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1; Transplantability-associated protein 1
Gene Aliases: MmTRA1a; MMTRA1B; Nor1; PLSCR1; Tra1; Tra1a; Tra1b; Tras1; Tras2
UniProt ID: (Human) O15162, (Rat) P58195, (Mouse) Q9JJ00
Entrez Gene ID: (Human) 5359, (Rat) 117540, (Mouse) 22038
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.