Novus Biologicals
Manufacturer Code:NBP300156UV
Catalog # NB064677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PLRP1 (AAH25784.1 Met1-Cys467). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1 EC 3.1.1.3 pancreatic lipase-related protein 1 PLRP1PL-RP1; Inactive pancreatic lipase-related protein 1; pancreatic lipase-related protein 1; PL-RP1
Gene Aliases: PLRP1; PNLIPRP1
UniProt ID: (Human) P54315
Entrez Gene ID: (Human) 5407
Molecular Function:
esterase
hydrolase
lipase
phospholipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.