Novus Biologicals
Manufacturer Code:NBP297702AF647
Catalog # NB046779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PLOD3 fragment. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate 5-dioxygenase 3 EC 1.14.11.4 LH3Lysyl hydroxylase 3 lysine hydroxylase 3 procollagen-lysine procollagen-lysine 2-oxoglutarate 5-dioxygenase 3; Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; Lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; Procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Gene Aliases: LH3; PLOD3
UniProt ID: (Human) O60568
Entrez Gene ID: (Human) 8985
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.