Abnova
Manufacturer Code:H00008985K
Catalog # 89-209-929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PLOD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PLOD3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; Lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; Procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Gene Aliases: LH3; PLOD3
UniProt ID: (Human) O60568
Entrez Gene ID: (Human) 8985
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.