Invitrogen
Manufacturer Code:PA542896
Catalog # PIPA542896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FXYD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2°-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FXYD domain containing ion transport regulator 1 MGC44983 PLM; FXYD domain-containing ion transport regulator 1; Phospholemman; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Aliases: FXYD1; PLM
UniProt ID: (Bovine) Q3SZX0, (Human) O00168, (Rat) O08589
Entrez Gene ID: (Bovine) 616139, (Human) 5348, (Dog) 476487, (Horse) 100630886, (Rat) 58971
Molecular Function: enzyme modulator ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.