Invitrogen
Manufacturer Code:PA537987
Catalog # PIPA537987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence SLDVPRSHGCAPTT from the internal region of PLK5 (aa309-322) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Inactive serine/threonine-protein kinase PLK5; PLK-5; PLK-5 PLK5P SgK384ps inactive serine/threonine-protein kinase PLK5 polo-like kinase 5 pseudogene; Polo-like kinase 5; polo-like kinase 5, pseudogene
Gene Aliases: FG060302; PLK-5; PLK5; PLK5P; SgK384ps
UniProt ID: (Human) Q496M5
Entrez Gene ID: (Human) 126520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.