Novus Biologicals
Manufacturer Code:NBP197306C
Catalog # NBP197306C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human PLK5 protein (between residues 200-250) [UniProt Q496M5] |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inactive serine/threonine-protein kinase PLK5; MGC118807 MGC118808 PLK5P polo-like kinase 5 polo-like kinase 5 pseudogene SgK384ps; PLK-5; Polo-like kinase 5; polo-like kinase 5, pseudogene
Gene Aliases: FG060302; PLK-5; PLK5; PLK5P; SgK384ps
UniProt ID: (Human) Q496M5
Entrez Gene ID: (Human) 126520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.