Novus Biologicals
Manufacturer Code:NB100894
Catalog # NB100894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence ATCIGEKIEDFKVGN corresponding to N-Terminus according to NP_055079.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11.21 PLK-4 polo-like kinase 4 (Drosophila) polo-like kinase 4SAKSTK18serine/threonine kinase 18 Sak serine/threonine protein kinase SAK Serine/threonine-protein kinase 18 serine/threonine-protein kinase PLK4 Serine/threonine-protein kinase Sak Snk akin kinase; PLK-4; Polo-like kinase 4; Serine/threonine-protein kinase 18; Serine/threonine-protein kinase PLK4; Serine/threonine-protein kinase Sak; Snk akin kinase
Gene Aliases: MCCRP2; PLK4; SAK; STK18
UniProt ID: (Human) O00444
Entrez Gene ID: (Human) 10733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.