Invitrogen
Manufacturer Code:PA579832
Catalog # PIPA579832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human PLK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSNK; PLK-2; polo-like kinase 1; Polo-like kinase 2; Serine/threonine-protein kinase PLK2; Serine/threonine-protein kinase PLK2 Polo-like kinase 2 PLK-2 hPlk2 Serine/threonine-protein kinase SNK hSNK Serum-inducible kinase PLK2 SNK; Serine/threonine-protein kinase SNK; Serum-inducible kinase
Gene Aliases: hPlk2; hSNK; PLK2; SNK
UniProt ID: (Human) Q9NYY3, (Mouse) P53351, (Rat) Q9R012
Entrez Gene ID: (Human) 10769, (Mouse) 20620, (Rat) 83722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.