Novus Biologicals
Manufacturer Code:NBP2842220.1ML
Catalog # NB032150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of PLK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 PLK-2 polo-like kinase 2EC 2.7.11.21 serine/threonine-protein kinase PLK2 Serine/threonine-protein kinase SNK Serum-inducible kinase SNKpolo-like kinase 2 (Drosophila); hSNK; PLK-2; Polo-like kinase 2; Serine/threonine-protein kinase PLK2; Serine/threonine-protein kinase SNK; Serum-inducible kinase
Gene Aliases: hPlk2; hSNK; PLK2; SNK
UniProt ID: (Human) Q9NYY3
Entrez Gene ID: (Human) 10769
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.