Novus Biologicals
Manufacturer Code:NBP222196
Catalog # NBP222196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C11 |
Immunogen | Purified recombinant fragment of human PLK1 (AA: 331-508) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cell cycle regulated protein kinase; cell cycle regulated protein kinase EC 2.7.11 EC 2.7.11.21 PLK PLK-1 polo (Drosophia)-like kinase polo like kinase polo-like kinase 1polo-like kinase (Drosophila) Serine/threonine-protein kinase 13 serine/threonine-protein kinase PLK1 STPK13; PLK-1; polo (Drosophia)-like kinase; polo like kinase; Polo-like kinase 1; Serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13
Gene Aliases: PLK; PLK1; STPK13
UniProt ID: (Human) P53350
Entrez Gene ID: (Human) 5347
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.