Invitrogen
Manufacturer Code:PA141319
Catalog # 01-675-656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 100-200 of human PLK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle regulated protein kinase; PLK-1; PLK1STPK14; polo (Drosophia)-like kinase; polo like kinase; Polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; Serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13
Gene Aliases: PLK; PLK1; STPK13
UniProt ID: (Bovine) Q2TA25, (Human) P53350, (Mouse) Q07832, (Rat) Q62673
Entrez Gene ID: (Pig) 396953, (Bovine) 538238, (Dog) 489971, (Human) 5347, (Mouse) 18817, (Rat) 25515
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.