Invitrogen
Manufacturer Code:PA5110764
Catalog # PIPA5110764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human PLGRKT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110007H22Rik 5033414D02Rik AD025 AI852040 C9orf46 MDS030 Plasminogen receptor (KT) Plg-R(KT) Plgrkt PLG-RKT RGD1306839; 5033414D02Rik; Plasminogen receptor (KT); plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); transmembrane protein C9orf46; transmembrane protein C9orf46 homolog
Gene Aliases: 1110007H22Rik; 5033414D02Rik; AD025; AI852040; C9orf46; MDS030; Plg-R(KT); PLG-RKT; PLGRKT; RGD1306839
UniProt ID: (Human) Q9HBL7, (Rat) D4ACN8, (Mouse) Q9D3P8
Entrez Gene ID: (Human) 55848, (Rat) 293888, (Mouse) 67759
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.