Novus Biologicals
Manufacturer Code:NB11096419
Catalog # NB11096419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 178/G10 |
Immunogen | Recombinant human PlGF-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D12S1900 PGFL placenta growth factor placental growth factor placental growth factor vascular endothelial growth factor-related protein PlGF PlGF-2 PLGFplacental growth factor-like SHGC-10760; Placenta growth factor; placental growth factor, vascular endothelial growth factor-related protein; PlGF
Gene Aliases: D12S1900; PGF; PGFL; PIGF; PLGF; PlGF-2; SHGC-10760
UniProt ID: (Human) P49763
Entrez Gene ID: (Human) 5228
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.