Abnova
Manufacturer Code:PAB5072
Catalog # 89-131-719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified mouse Plg. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native Plg.
For Research Use Only
Protein Aliases: Activation peptide; AI649309; Angiostatin; Pg; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen
Gene Aliases: AI649309; Pg; Plg
UniProt ID: (Mouse) P20918
Entrez Gene ID: (Mouse) 18815
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.