Abnova
Manufacturer Code:PAB29130
Catalog # 89-130-608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Dog |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified dog PLG with Freund's adjuvant. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against native dog PLG.
For Research Use Only
Protein Aliases: angiostatin fragment
Entrez Gene ID: (Dog) 403602
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.