Abnova
Manufacturer Code:PAB16862
Catalog # 89-118-677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 17 amino acids of human PLEKHM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PLEKHM2.
For Research Use Only
Protein Aliases: KIAA0842; novel RUN and PH domain-containing protein; PH domain-containing family M member 2; pleckstrin homology domain containing, family M (with RUN domain) member 2; Pleckstrin homology domain-containing family M member 2; RP11-169K16.1; Salmonella-induced filaments A and kinesin-interacting protein; SifA (Salmonella-induced filaments A) and kinesin-interacting protein; SifA and kinesin-interacting protein; SKIP
Gene Aliases: KIAA0842; PLEKHM2; SKIP
UniProt ID: (Human) Q8IWE5
Entrez Gene ID: (Human) 23207
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.