Novus Biologicals
Manufacturer Code:NBP170676
Catalog # NBP170676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PLEKHA9(pleckstrin homology domain containing family A member 9) The peptide sequence was selected from the N terminal of PLEKHA9. Peptide sequence VGTLLKSTCNTFLKTLEECMQIANAAFTSELLYHTPPGSPQLAMLKSSKM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PLEKHA8P1 pleckstrin homology domain containing family A member 8 pseudogene 1
Gene Aliases: PLEKHA9
Entrez Gene ID: (Human) 51054
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.