Proteintech
Manufacturer Code:175681AP150UL
Catalog # 50-173-1652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLEKHA9 Fusion Protein Ag11772 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAPP-2; PH domain-containing family A member 8; Phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; Pleckstrin homology domain-containing family A member 8; PLEKHA9 Protein PLEKHA9
Gene Aliases: 6330400G01; AA517676; BC052360; FAPP-2; Fapp2; Plekha8; PLEKHA9; RGD1563940
UniProt ID: (Mouse) Q80W71, (Rat) D3ZY60
Entrez Gene ID: (Human) 51054, (Mouse) 231999, (Rat) 500132
Molecular Function:
nucleic acid binding
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.