Proteintech
Manufacturer Code:154101AP150UL
Catalog # 50-173-0207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLEKHA8 Fusion Protein Ag7677 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAPP 2 FAPP2 hFAPP2 PLEKHA8; FAPP-2; PH domain-containing family A member 8; Phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; Pleckstrin homology domain-containing family A member 8; Serologically defined breast cancer antigen NY-BR-86
Gene Aliases: 6330400G01; AA517676; BC052360; FAPP-2; FAPP2; PLEKHA8; RGD1563940
UniProt ID: (Human) Q96JA3, (Mouse) Q80W71, (Rat) D3ZY60
Entrez Gene ID: (Human) 84725, (Mouse) 231999, (Rat) 500132
Molecular Function: nucleic acid binding transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.