Proteintech
Manufacturer Code:268301AP150UL
Catalog # 50-173-5471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLEKHA1 Fusion Protein Ag25285 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PH domain-containing family A member 1; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1; Pleckstrin homology domain-containing family A member 1; PLEKHA1 TAPP 1 TAPP1; Tandem PH domain-containing protein 1; TAPP-1
Gene Aliases: PLEKHA1; TAPP1
UniProt ID: (Human) Q9HB21
Entrez Gene ID: (Human) 59338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.